Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENP-E Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501561374
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-1374 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1374 Supplier Bethyl Laboratories, Inc Supplier No. A301943AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The kinetophore is a macromolecular structure that is associated with the centromeres of chromosomes and is responsible for establishing and maintaining the connections with the microtubules of the mitotic spindle. The proteins that reside within the chromosomal domain fall into two broad classes: those that associate with the centromere throughout the cell cycle and those that are found at the centromere in a cell cycle specific manner. Members of the first class include CENP-A, CENP-B, and CENP-C. The members of the second class include several ubiquitous cellular proteins such as calmodulin, tubulin, dynein and p34 cdc2 and also CENP-E and CENP-F. CENP-F appears to be the earliest member to interact with the centromere-kinetochore complex. CENP-F also shows a cell-cycle distribution that is both temporally regulated and diverse in terms of the structural components it localizes to and therefore may play a role in several mitotic events.

Specifications

Antigen CENP-E
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CENPE
Gene Accession No. Q02224
Gene Alias centromere protein E
Gene Symbols CENPE
Host Species Rabbit
Immunogen Between 2500 and 2550
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1062
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.