Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENP-EMouse anti-Human, Clone: 1H12, Rockland™
SDP

Catalog No. 501990541
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0541 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0541 Supplier Rockland Immunochemicals Supplier No. 200301901
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against human CENP-E protein. The product was purified from tissue culture supernatant by chromatography. This antibody reacts with CENP-E from human cells. Reactivity against homologues from other sources is not known.

14-3-3 sigma (stratifin, SFN, epithelial cell marker protein 1) is a member of a highly conserved family of 14-3-3 proteins that are present in all eukaryotic organisms. There are 7 known mammalian 14-3-3 isoforms epsilon, beta, zeta, gamma, theta, tau and they play important roles in many biological activities by binding to and altering the subcellular localization and/or stability of key molecules in various signaling cascades. 14-3-3 sigma was originally characterized as a human mammary epithelium marker and later rediscovered as an important molecule for cell cycle checkpoint regulation. Recently it has been reported that 14-3-3 sigma may serve as a prognosis marker predicting survival of pancreatic cancer patient treatment. 14-3-3 sigmais the only 14-3-3 isoform induced by the tumor suppressor protein p53, in response to γ-irradiation and other DNA-damaging agents. 14-3-3 sigmais a p53-regulated inhibitor of G2/M progression and acts as a tumor suppressor gene that is inactivated by methylation of its 5' CpG islands in epithelial tumor cells. Two isoforms of human 14-3-3 sigma are produced by alternative splicing.

Specifications

Antigen CENP-E
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene CENPE
Gene Accession No. Q02224
Gene Alias CENPE; CENP-E; Centromere autoantigen E (312kD); Centromere protein E; centromere protein E, 312kDa; Centromere-associated protein E; KIF10; kinesin family member 10; Kinesin-7; Kinesin-related protein CENPE; MCPH13; PPP1R61; protein phosphatase 1, regulatory subunit 61
Gene Symbols CENPE
Host Species Mouse
Immunogen This protein A purified monoclonal antibody was produced by repeated immunizations with a full length recombinant protein corresponding to human CENP-E protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1062
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.