Learn More
CENP-F/Mitosin Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301611A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, acetone fixation is recommended.
This gene encodes a protein that associates with the centromere-kinetochore complex. The protein is a component of the nuclear matrix during the G2 phase of interphase. In late G2 the protein associates with the kinetochore and maintains this association through early anaphase. It localizes to the spindle midzone and the intracellular bridge in late anaphase and telophase, respectively, and is thought to be subsequently degraded. The localization of this protein suggests that it may play a role in chromosome segregation during mitotis. It is thought to form either a homodimer or heterodimer. Autoantibodies against this protein have been found in patients with cancer or graft versus host disease.Specifications
| CENP-F/Mitosin | |
| Polyclonal | |
| Unconjugated | |
| CENPF | |
| RP11-262H5.1, CENF, PRO1779, hcp-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1063 | |
| 4° C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| P49454 | |
| CENPF | |
| Between 1800 and 1850 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.