Learn More
CENP-J Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302986AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee
This gene encodes a protein that belongs to the centromere protein family. During cell division, this protein plays a structural role in the maintece of centrosome integrity and normal spindle morphology, and it is involved in microtubule disassembly at the centrosome. This protein can function as a transcriptional coactivator in the Stat5 signaling pathway, and also as a coactivator of NF-kappaB-mediated transcription, likely via its interaction with the coactivator p300/CREB-binding protein. Mutations in this gene are associated with primary autosomal recessive microcephaly, a disorder characterized by severely reduced brain size and mental retardation.Specifications
CENP-J | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9HC77 | |
CENPJ | |
Between 675 and 725 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CENPJ | |
BM032, CENP-J, centrosomal P4.1-associated protein, CPAP, LAG-3-associated protein, LAP, LIP1, LYST-interacting protein 1, LYST-interacting protein LIP1, LYST-interacting protein LIP7, MCPH6, microcephaly, primary autosomal recessive 6, Sas-4, SASS4, SCKL4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55835 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.