Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPA Antibody (5A7-2E11), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212892R
Description
CENPA Monoclonal antibody specifically detects CENPA in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CENPA | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
5A7-2E11 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Centromere autoantigen A, centromere protein A (17kD), centromere protein ACENP-Acentromere protein A, 17kDa, histone H3-like centromeric protein A | |
Synthetic peptide corresponding to a portion of human CENP-A | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication | |
1058 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction