Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313389
This item is not returnable.
View return policy
Description
CENPA Polyclonal antibody specifically detects CENPA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CENPA | |
| Polyclonal | |
| Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Centromere autoantigen A, centromere protein A (17kD), centromere protein ACENP-Acentromere protein A, 17kDa, histone H3-like centromeric protein A | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human CENPA. The exact sequence is proprietary. | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication | |
| 1058 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction