Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPA Monoclonal Antibody (5A7-2E11), PE
GREENER_CHOICE

Catalog No. PIMA545300
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545300 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA545300 Supplier Invitrogen™ Supplier No. MA545300
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

1 μg/mL of MA5-45300 was sufficient for detection of CENPA in 20 μg of U2OS cell lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 18kDa. Recognizes Human CENP-A.

Centromeres are the differentiated chromosomal domains that specify the mitotic behavior of chromosomes. CENPA encodes a centromere protein which contains a histone H3 related histone fold domain that is required for targeting to the centromere. CENPA is proposed to be a component of a modified nucleosome or nucleosome-like structure in which it replaces 1 or both copies of conventional histone H3 in the (H3-H4)2 tetrameric core of the nucleosome particle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPA
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5A7-2E11
Concentration 1 mg/mL
Conjugate PE
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative
Gene Cenpa
Gene Accession No. P49450
Gene Alias CenH3; CENP A; CENPA; CENP-A; Centromere autoantigen A; centromere protein A; centromere protein A, 17kDa; centromere-specific histone; ggCENP-A; histone H3-like centromeric protein A; RGD1563607
Gene Symbols Cenpa
Host Species Mouse
Immunogen Synthetic peptide corresponding to a portion of human CENP-A.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1058
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.