Learn More
Invitrogen™ CENPA Monoclonal Antibody (5A7-2E11)

Description
1 μg/mL of MA5-27710 was sufficient for detection of CENPA in 20 μg of U2OS cell lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 18kDa. Recognizes Human CENP-A.
Specifications
Specifications
| Antigen | CENPA |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5A7-2E11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 |
| Gene | Cenpa |
| Gene Accession No. | P49450 |
| Gene Alias | CenH3; CENP A; CENPA; CENP-A; Centromere autoantigen A; centromere protein A; centromere protein A, 17kDa; centromere-specific histone; ggCENP-A; histone H3-like centromeric protein A; RGD1563607 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.