Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPBD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566334 Supplier Invitrogen™ Supplier No. PA566334
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERPTDATVIPS AKRERKAITL DLKLEVLRRF EAGEKLSQIA KALDLAISTV Highest antigen sequence identity to the following orthologs - mouse 31%, rat 31%.

CENPBD1 gene ontology annotations related to this gene include DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPBD1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CENPBD1
Gene Accession No. B2RD01
Gene Alias CENPB DNA-binding domain containing 1; CENPB DNA-binding domain-containing protein 1; CENPB DNA-binding domains containing 1; CENPBD1
Gene Symbols CENPBD1
Host Species Rabbit
Immunogen Recombinant Human CENPBD1. Recombinant protein control fragment (Product #RP-107014).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92806
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.