Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPC Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CENPC |
---|---|
Concentration | 0.35 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9317
![]() ![]() |
Proteintech Group Inc
142501AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Centromere protein C 1 is a centromere autoantigen and a component of the inner kinetochore plate. The protein is required for maintaining proper kinetochore size and a timely transition to anaphase. A putative pseudogene exists on chromosome 12.Specifications
CENPC | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q03188 | |
1060, 12617 | |
CENPC Fusion Protein Ag5523 | |
Primary | |
-20°C | |
CENPC |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CENP C, CENP C 1, CENPC, CENPC1, Centromere autoantigen C, centromere protein C 1, hcp 4, ICEN7, MIF2 | |
CENPC, Cenpc1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title