Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983877
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3877 Supplier Bioss Supplier No. BS15555R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Centromere protein C 1 is a centromere autoantigen and a component of the inner kinetochore plate. The protein is required for maintaining proper kinetochore size and a timely transition to anaphase. A putative pseudogene exists on chromosome 12.

Specifications

Antigen CENPC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CENPC
Gene Accession No. Q03188
Gene Alias Cenpc; CENP-C; CENP-C 1; CENPC1; Centromere autoantigen C; centromere autoantigen C1; Centromere protein C; CENTROMERE PROTEIN C (CENP-C) (CENTROMERE AUTOANTIGEN C); Centromere protein C 1; centromere protein C1; hcp-4; ICEN7; Interphase centromere complex protein 7; MIF2
Gene Symbols CENPC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CENPC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1060
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.