Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    CENPE Antibody, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106506
Description
CENPE Polyclonal specifically detects CENPE in Human samples. It is validated for Immunoprecipitation.Specifications
| CENPE | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| CENP-E, Centromere autoantigen E (312kD), Centromere protein E, centromere protein E (312kD), centromere protein E, 312kDa, centromere-associated protein E, KIF10, kinesin family member 10, Kinesin-related protein CENPE | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG | 
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 1:10-1:500 | |
| NP_001804.2 | |
| CENPE | |
| The immunogen recognized by this antibody maps to a region between residue 2500 and 2550 of human centromere protein E using the numbering given in entry NP_001804.2 (GeneID 1062). | |
| 100 μL | |
| Cell Cycle and Replication | |
| 1062 | |
| Store at 4C. Do not freeze. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            For Research Use Only
Spot an opportunity for improvement?Share a Content Correction