Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPE Monoclonal Antibody (1H12)
GREENER_CHOICE

Catalog No. p-7171697
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-178 100 μL
PIMA518025 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-675-178 Supplier Invitrogen™ Supplier No. MA15758
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This target has a predicted molecular weight of 312 kDa and is only suitable for Western blot if IP is performed first.

14-3-3 sigma (stratifin, SFN, epithelial cell marker protein 1) is a member of a highly conserved family of 14-3-3 proteins that are present in all eukaryotic organisms. There are 7 known mammalian 14-3-3 isoforms epsilon, beta, zeta, gamma, theta, tau and they play important roles in many biological activities by binding to and altering the subcellular localization and/or stability of key molecules in various signaling cascades. 14-3-3 sigma was originally characterized as a human mammary epithelium marker and later rediscovered as an important molecule for cell cycle checkpoint regulation. Recently it has been reported that 14-3-3 sigma may serve as a prognosis marker predicting survival of pancreatic cancer patient treatment. 14-3-3 sigmais the only 14-3-3 isoform induced by the tumor suppressor protein p53, in response to <FONT face=Symbol>g</FONT>-irradiation and other DNA-damaging agents. 14-3-3 sigmais a p53-regulated inhibitor of G2/M progression and acts as a tumor suppressor gene that is inactivated by methylation of its 5' CpG islands in epithelial tumor cells. Two isoforms of human 14-3-3 sigma are produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPE
Applications Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene CENPE
Gene Accession No. Q02224
Gene Alias 312kDa; AU019344; BC049989; C530022J18; Cenpe; CENP-E; centromere autoantigen E; Centromere autoantigen E (312kD); centromere protein E; centromere protein E, 312kDa; centromere-associated protein E; centromeric protein E; KIF10; kinesin 10; kinesin family member 10; kinesin superfamily protein 10; Kinesin-7; Kinesin-related protein CENPE; MCPH13; Motor domain of KIF10; N-7 kinesin; PPP1R61; protein phosphatase 1, regulatory subunit 61
Gene Symbols CENPE
Host Species Mouse
Immunogen Recombinant full length CENP-E protein (human) from SP2/0 cells.
Purification Method Ion-exchange chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1062
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.