Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559939 Supplier Invitrogen™ Supplier No. PA559939
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKIENLTRML VTSSSLTLQQ ELKAKRKRRV TWCLGKINKM KNSNYADQFN IPTNITTKTH KLSINLLREI DESVCSESDV FSNTLDTLSE IEWNPATKLL NQENIESELN SLRADYDN Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 60%.

14-3-3 sigma (stratifin, SFN, epithelial cell marker protein 1) is a member of a highly conserved family of 14-3-3 proteins that are present in all eukaryotic organisms. There are 7 known mammalian 14-3-3 isoforms epsilon, beta, zeta, gamma, theta, tau and they play important roles in many biological activities by binding to and altering the subcellular localization and/or stability of key molecules in various signaling cascades. 14-3-3 sigma was originally characterized as a human mammary epithelium marker and later rediscovered as an important molecule for cell cycle checkpoint regulation. Recently it has been reported that 14-3-3 sigma may serve as a prognosis marker predicting survival of pancreatic cancer patient treatment. 14-3-3 sigmais the only 14-3-3 isoform induced by the tumor suppressor protein p53, in response to <FONT face=Symbol>g</FONT>-irradiation and other DNA-damaging agents. 14-3-3 sigmais a p53-regulated inhibitor of G2/M progression and acts as a tumor suppressor gene that is inactivated by methylation of its 5' CpG islands in epithelial tumor cells. Two isoforms of human 14-3-3 sigma are produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPE
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CENPE
Gene Accession No. Q02224
Gene Alias 312kDa; AU019344; BC049989; C530022J18; Cenpe; CENP-E; centromere autoantigen E; Centromere autoantigen E (312kD); centromere protein E; centromere protein E, 312kDa; centromere-associated protein E; centromeric protein E; KIF10; kinesin 10; kinesin family member 10; kinesin superfamily protein 10; Kinesin-7; Kinesin-related protein CENPE; MCPH13; Motor domain of KIF10; N-7 kinesin; PPP1R61; protein phosphatase 1, regulatory subunit 61
Gene Symbols CENPE
Host Species Rabbit
Immunogen Recombinant protein fragment of Human CENPE (Product #RP-89483)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1062
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.