Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPF Monoclonal Antibody (7F11.2)
GREENER_CHOICE

Catalog No. PIMA531600
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531600 Supplier Invitrogen™ Supplier No. MA531600
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Positive Control: HeLa.

This gene encodes a protein that associates with the centromere-kinetochore complex. The protein is a component of the nuclear matrix during the G2 phase of interphase. In late G2 the protein associates with the kinetochore and maintains this association through early anaphase. It localizes to the spindle midzone and the intracellular bridge in late anaphase and telophase, respectively, and is thought to be subsequently degraded. The localization of this protein suggests that it may play a role in chromosome segregation during mitosis. It is thought to form either a homodimer or heterodimer. Autoantibodies against this protein have been found in patients with cancer or graft versus host disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPF
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 7F11.2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2
Gene CENPF
Gene Accession No. P49454
Gene Alias 6530404A22Rik; AH antigen; AI325968; cell-cycle-dependent 350K nuclear protein; CENF; CENPF; CENP-F; CENP-F kinetochore protein; centromere autoantigen F; centromere protein F; centromere protein F, (mitosin); centromere protein F, 350/400ka (mitosin); centromere protein F, 350/400kDa (mitosin); CILD31; hcp-1; im:7140452; im:7144238; im:7154719; im:7156761; kinetochore protein CENPF; Lek1; leucine, glutamic acid, lysine family 1 protein; mitosin; PRO1779; RP11-262H5.1; si:ch211-152h14.2; STROMS
Gene Symbols CENPF
Host Species Mouse
Immunogen Protein fragment expressed in E. coli corresponding to amino acids 1759-2093.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1063
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.