Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100716 Supplier Invitrogen™ Supplier No. PA5100716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CENPF.

This gene encodes a protein that associates with the centromere-kinetochore complex. The protein is a component of the nuclear matrix during the G2 phase of interphase. In late G2 the protein associates with the kinetochore and maintains this association through early anaphase. It localizes to the spindle midzone and the intracellular bridge in late anaphase and telophase, respectively, and is thought to be subsequently degraded. The localization of this protein suggests that it may play a role in chromosome segregation during mitosis. It is thought to form either a homodimer or heterodimer. Autoantibodies against this protein have been found in patients with cancer or graft versus host disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CENPF
Gene Accession No. P49454
Gene Alias 6530404A22Rik; AH antigen; AI325968; cell-cycle-dependent 350K nuclear protein; CENF; CENPF; CENP-F; CENP-F kinetochore protein; centromere autoantigen F; centromere protein F; centromere protein F, (mitosin); centromere protein F, 350/400ka (mitosin); centromere protein F, 350/400kDa (mitosin); CILD31; hcp-1; im:7140452; im:7144238; im:7154719; im:7156761; kinetochore protein CENPF; Lek1; leucine, glutamic acid, lysine family 1 protein; mitosin; PRO1779; RP11-262H5.1; si:ch211-152h14.2; STROMS
Gene Symbols CENPF
Host Species Rabbit
Immunogen A synthesized peptide derived from human CENPF(Accession P49454), corresponding to amino acid residues 3072-3086.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1063, 108000
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.