Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPH Antibody (OTI2G4), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272163AF750
Description
CENPH Monoclonal specifically detects CENPH in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
CENPH | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CENPH | |
Full length recombinant protein of human CENPH (NP_075060) produced in HEK293T cells. | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI2G4 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
CENP-H, centromere protein H, ICEN35, Interphase centromere complex protein 35, kinetochore protein CENP-H, NNF1, PMF1 | |
Mouse | |
0.1 mL | |
64946 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction