Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPH Antibody (OTI2G4), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272163MFV500
Description
CENPH Monoclonal antibody specifically detects CENPH in Human samples. It is validated for Flow CytometrySpecifications
CENPH | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI2G4 | |
Flow Cytometry | |
CENP-H, centromere protein H, ICEN35, Interphase centromere complex protein 35, kinetochore protein CENP-H, NNF1, PMF1 | |
Full length human recombinant protein of human CENPH(NP_075060) produced in HEK293T cell. | |
0.1 mL | |
Cell Cycle and Replication | |
64946 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction