Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPH Monoclonal Antibody (OTI1A6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503874S


Description
CENPH Monoclonal antibody specifically detects CENPH in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
CENPH | |
Monoclonal | |
0.73 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9H3R5 | |
Mouse | |
Affinity chromatography | |
RUO | |
64946 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI1A6 | |
Unconjugated | |
CENPH | |
1700021I11Rik; 2410018A12Rik; 2610042E16Rik; 2810046K12Rik; AU044255; CENPH; CENP-H; centromere autoantigen H; centromere protein H; Enp; ICEN35; Interphase centromere complex protein 35; kinetochore protein CENP-H; LOC681185; similar to centromere autoantigen H | |
Full length human recombinant protein of human CENPH produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction