Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPI Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156364
Description
CENPI Polyclonal specifically detects CENPI in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CENPI | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CENP-IFSH primary response (LRPR1, rat) homolog 1, centromere protein I, Follicle-stimulating hormone primary response protein, FSH primary response 1, FSH primary response protein 1, FSHPRH1Mis6, ICEN19, Interphase centromere complex protein 19, Leucine-rich primary response protein 1, LRPR1FSH primary response (LRPR1 homolog, rat) 1, Mis6 | |
Rabbit | |
87 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%. | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence | |
Q92674 | |
CENPI | |
Synthetic peptides corresponding to CENPI(centromere protein I) The peptide sequence was selected from the N terminal of CENPI. Peptide sequence SPQKRVKNVQAQNRTSQGSSSFQTTLSAWKVKQDPSNSKNISKHGQNNPV. | |
Affinity purified | |
RUO | |
2491 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction