Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100717
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100717 Supplier Invitrogen™ Supplier No. PA5100717
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CENPI.

A novel gene containing SNF2, PHD-finger, RING-finger, helicase, and linker histone domains termed SHPRH, is a possible candidate for a tumor suppressor gene. These domains are characteristic of several DNA repair proteins, transcription factors, and helicases. Mapping of this gene to 6q24, a region reported to contain a tumor suppressor locus suggests a role in tumor suppression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPI
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cenpi
Gene Accession No. Q92674
Gene Alias 2610103K11Rik; AA450458; AU024614; BC006883; CENPI; CENP-I; Centromere protein I; D230017O13Rik; E130018M05; follicle-stimulating hormone primary response protein; FSH primary response 1; FSH primary response protein 1; FSHPRH1; ICEN19; Interphase centromere complex protein 19; leucine-rich primary response protein 1; LRPR1; Mis6
Gene Symbols Cenpi
Host Species Rabbit
Immunogen A synthesized peptide derived from human CENPI(Accession Q92674), corresponding to amino acid residues S306-C356.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2491
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.