Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPJ Monoclonal Antibody (OTI3G8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811852

Description
This gene encodes a protein that belongs to the centromere protein family. During cell division, this protein plays a structural role in the maintece of centrosome integrity and normal spindle morphology, and it is involved in microtubule disassembly at the centrosome. This protein can function as a transcriptional coactivator in the Stat5 signaling pathway, and also as a coactivator of NF-kappaB-mediated transcription, likely via its interaction with the coactivator p300/CREB-binding protein. Mutations in this gene are associated with primary autosomal recessive microcephaly, a disorder characterized by severely reduced brain size and mental retardation.Specifications
CENPJ | |
Monoclonal | |
Unconjugated | |
CENPJ | |
BM032, CENP-J, CPAP, LAP, LIP1, MCPH6, SASS4, SCKL4, Sas-4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55835 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI3G8 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9HC77 | |
CENPJ | |
Human recombit protein fragment corresponding to amino acids 1087-1338 of human CENPJ produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction