Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPJ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100718 Supplier Invitrogen™ Supplier No. PA5100718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CENPJ.

This gene encodes a protein that belongs to the centromere protein family. During cell division, this protein plays a structural role in the maintenance of centrosome integrity and normal spindle morphology, and it is involved in microtubule disassembly at the centrosome. This protein can function as a transcriptional coactivator in the Stat5 signaling pathway, and also as a coactivator of NF-kappaB-mediated transcription, likely via its interaction with the coactivator p300/CREB-binding protein. Mutations in this gene are associated with primary autosomal recessive microcephaly, a disorder characterized by severely reduced brain size and mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPJ
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CENPJ
Gene Accession No. Q569L8, Q9HC77
Gene Alias 4932437H03Rik; BM032; Cenpj; CENP-J; Centromere protein J; centrosomal P4.1-associated protein; CPAP; Gm81; LAG-3-associated protein; LAP; LIP1; LYST-interacting protein 1; LYST-interacting protein LIP1; LYST-interacting protein LIP7; MCPH6; microcephaly; Sas4; Sas-4; SASS4; SCKL4
Gene Symbols CENPJ
Host Species Rabbit
Immunogen A synthesized peptide derived from human CENPJ(Accession Q9HC77), corresponding to amino acid residues F1265-E1315.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219103, 55835
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.