Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CENPJ Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550377
Description
The antibody detects endogenous levels of total CENPJ protein.
This gene encodes a protein that belongs to the centromere protein family. During cell division, this protein plays a structural role in the maintenance of centrosome integrity and normal spindle morphology, and it is involved in microtubule disassembly at the centrosome. This protein can function as a transcriptional coactivator in the Stat5 signaling pathway, and also as a coactivator of NF-kappaB-mediated transcription, likely via its interaction with the coactivator p300/CREB-binding protein. Mutations in this gene are associated with primary autosomal recessive microcephaly, a disorder characterized by severely reduced brain size and mental retardation.
Specifications
CENPJ | |
Polyclonal | |
Unconjugated | |
CENPJ | |
4932437H03Rik; BM032; Cenpj; CENP-J; Centromere protein J; centrosomal P4.1-associated protein; CPAP; Gm81; LAG-3-associated protein; LAP; LIP1; LYST-interacting protein 1; LYST-interacting protein LIP1; LYST-interacting protein LIP7; MCPH6; microcephaly; Sas4; Sas-4; SASS4; SCKL4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55835 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9HC77 | |
CENPJ | |
Fusion protein corresponding to residues near the C terminal of human centromere protein J. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction