Learn More
Invitrogen™ CENPK Polyclonal Antibody

Description
CENPK Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | CENPK |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CENPK |
| Gene Accession No. | Q9BS16, Q9ESN5 |
| Gene Alias | AF5alpha; B130045K24Rik; BB232189; C530004N04Rik; CENPK; Cenp-K; Centromere protein K; FKSG14; ICEN37; Interphase centromere complex protein 37; leucine zipper protein FKSG14; p33; Protein AF-5alpha; Solt; Solzt; SoxLZ/Sox6 leucine zipper binding protein in testis; SoxLZ/Sox6 leucine zipper-binding protein in testis; SoxLZ/Sox6-binding protein Solt |
| Gene Symbols | CENPK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.