Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597578
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597578 Supplier Invitrogen™ Supplier No. PA597578
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CENPK Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA

CENPK is a subunit of a CENPH -CENPI -associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPK
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CENPK
Gene Accession No. Q9BS16
Gene Alias AF5alpha; B130045K24Rik; BB232189; C530004N04Rik; CENPK; Cenp-K; Centromere protein K; FKSG14; ICEN37; Interphase centromere complex protein 37; leucine zipper protein FKSG14; p33; Protein AF-5alpha; Solt; Solzt; SoxLZ/Sox6 leucine zipper binding protein in testis; SoxLZ/Sox6 leucine zipper-binding protein in testis; SoxLZ/Sox6-binding protein Solt
Gene Symbols CENPK
Host Species Rabbit
Immunogen Recombinant Human Centromere protein K protein (1-269AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64105
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.