Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CENPL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100719
Description
Antibody detects endogenous levels of total CENPL.
CENPL is a subunit of a CENPH-CENPI-associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.
Specifications
CENPL | |
Polyclonal | |
Unconjugated | |
CENPL | |
2610300B10Rik; AW121806; AW550697; C1orf155; CENPL; CENP-L; Centromere protein L; dJ383J4.3; ICEN33; Interphase centromere complex protein 33; RGD1311868; RP3-383J4.1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
70454, 91687 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q3U3S3, Q8N0S6 | |
CENPL | |
A synthesized peptide derived from human CENPL(Accession Q8N0S6), corresponding to amino acid residues W241-F291. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction