Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPL Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13836R

Description
CENPL is a subunit of a CENPH-CENPI-associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.Specifications
CENPL | |
Polyclonal | |
Unconjugated | |
CENPL | |
2610300B10Rik; AW121806; AW550697; C1orf155; Cenpl; CENP-L; Centromere protein L; dJ383J4.3; ICEN33; Interphase centromere complex protein 33; RGD1311868; RP3-383J4.1 | |
Rabbit | |
Protein A | |
RUO | |
91687 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH | |
Q8N0S6 | |
CENPL | |
KLH conjugated synthetic peptide derived from human CENPL. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction