Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPM Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | CENPM |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1543620
![]() |
Novus Biologicals
NBP15436120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP154361
![]() |
Novus Biologicals
NBP154361 |
100 μL |
Each for $487.50
|
|
|||||
Description
CENPM Polyclonal specifically detects CENPM in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CENPM | |
Polyclonal | |
Rabbit | |
Q9NSP4 | |
79019 | |
Synthetic peptides corresponding to CENPM(centromere protein M) The peptide sequence was selected from the middle region of CENPM. Peptide sequence CDLEVEGFRATMAQRLVRVLQICAGHVPGVSALNLLSLLRSSEGPSLEDL. | |
Primary |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
bK250D10.2, C22orf18PANE1MGC861, CENP-Mchromosome 22 open reading frame 18, centromere protein M, ICEN39, Interphase centromere complex protein 39, Pane1, proliferation associated nuclear element 1, Proliferation-associated nuclear element protein 1 | |
CENPM | |
IgG | |
20 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title