Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CENPM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542854
Description
Peptide sequence: CDLEVEGFRA TMAQRLVRVL QICAGHVPGV SALNLLSLLR SSEGPSLEDL Sequence homology: Cow: 79%; Dog: 79%; Horse: 93%; Human: 100%; Pig: 93%; Rabbit: 93%.
The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA replaces histone H3. CENPM is an additional factor required for centromere assembly.The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). CENPM is an additional factor required for centromere assembly (Foltz et al., 2006 [PubMed 16622419]).[supplied by OMIM].
Specifications
CENPM | |
Polyclonal | |
Unconjugated | |
CENPM | |
2610019I03Rik; AI853711; bK250D10.2; C22orf18; Cenpm; CENP-M; Centromere protein M; hypothetical protein LOC615953; ICEN39; Interphase centromere complex protein 39; MGC861; PANE1; proliferation associated nuclear element 1; proliferation-associated nuclear element protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79019 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NSP4 | |
CENPM | |
Synthetic peptide directed towards the middle region of human CENPM. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction