Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566020 Supplier Invitrogen™ Supplier No. PA566020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TEESLRHVDAS FFLGKVCFLA TGAGRESHCS IHRHTVVKLA HTYQSPLLYC DLEVEG Highest antigen sequence identity to the following orthologs - mouse 75%, rat 79%.

The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA replaces histone H3. CENPM is an additional factor required for centromere assembly.The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). CENPM is an additional factor required for centromere assembly (Foltz et al., 2006 [PubMed 16622419]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPM
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CENPM
Gene Accession No. Q9NSP4
Gene Alias 2610019I03Rik; AI853711; bK250D10.2; C22orf18; Cenpm; CENP-M; Centromere protein M; hypothetical protein LOC615953; ICEN39; Interphase centromere complex protein 39; MGC861; PANE1; proliferation associated nuclear element 1; proliferation-associated nuclear element protein 1
Gene Symbols CENPM
Host Species Rabbit
Immunogen Recombinant Human CENPM. Recombinant protein control fragment (Product #RP-106704).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.