Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544734 Supplier Invitrogen™ Supplier No. PA544734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SFKKILQRAL KNVTVSFRET EENAVWIRIA WGTQYTKPNQ YKPTYVVYYS Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Human: 100%; Mouse: 79%; Rabbit: 100%; Rat: 79%.

The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). CENPN is an additional factor required for centromere assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPN
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Cenpn
Gene Accession No. Q96H22
Gene Alias 2610510J17Rik; AI426416; ATM interactor; centromere protein N; AW545024; BM039; BM-309; C16orf60; Cenpn; CENP-N; Centromere protein N; Da2-27; Da2-4; FLJ13607; FLJ22660; ICEN32; Interphase centromere complex protein 32; RGD1310953; Unknown (protein for MGC:133728)
Gene Symbols Cenpn
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CENPN.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55839
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.