Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CENPN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565747
Description
Immunogen sequence: DETVAEFIKRT ILKIPMNELT TILKAWDFLS ENQLQTVNFR QRKESVVQHL IHLCEEKRAS ISDAALLDII YMQFHQHQKV WEV Highest antigen sequence identity to the following orthologs - mouse 63%, rat 63%.
The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). CENPN is an additional factor required for centromere assembly.
Specifications
CENPN | |
Polyclonal | |
Unconjugated | |
Cenpn | |
2610510J17Rik; AI426416; ATM interactor; centromere protein N; AW545024; BM039; BM-309; C16orf60; Cenpn; CENP-N; Centromere protein N; Da2-27; Da2-4; FLJ13607; FLJ22660; ICEN32; Interphase centromere complex protein 32; RGD1310953; Unknown (protein for MGC:133728) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55839 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96H22 | |
Cenpn | |
Recombinant Human CENPN. Recombinant protein control fragment (Product #RP-106423). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction