Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPO Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176240PCP
Description
CENPO Polyclonal antibody specifically detects CENPO in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
CENPO | |
Polyclonal | |
Western Blot, ELISA | |
CENP-OMGC11266, centromere protein O, centromeric protein O, ICEN36, ICEN-36, Interphase centromere complex protein 36, MCM21R | |
Antibody was raised against a 16 amino acid synthetic peptide from near the center of human CENPO. The immunogen is located within amino acids 170 - 220 of CENPO. Amino Acid Squence: AYSGRKYQADRLQSDF | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
PerCP | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
79172 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction