Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPQ Antibody (OTI1C5), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271991AF750
Description
CENPQ Monoclonal specifically detects CENPQ in Human samples. It is validated for Western Blot.Specifications
CENPQ | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CENPQ | |
Full length recombinant protein of human CENPQ (NP_060602) produced in E.coli. | |
DNA replication Transcription Translation and Splicing | |
55166 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1C5 | |
Western Blot | |
C6orf139, CENP-QFLJ10545, centromere protein Q, chromosome 6 open reading frame 139 | |
Mouse | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction