Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPQ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109512 Supplier Invitrogen™ Supplier No. PA5109512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEDLTNVSSL LNMERARDKA NEEGLALLQE EIDKMVETTE LMTGNIQSLK NKIQILASEV EEEEERVKQM HQINSSGVLS LPELSQKTLK APTLQKEILA LIPNQNALLK DLDILHNSSQ MKSMSTFIEE AYKKLDAS.

CENPQ is a subunit of a CENPH-CENPI-associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPQ
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CENPQ
Gene Accession No. Q7L2Z9
Gene Alias C6orf139; CENPQ; CENP-Q; Centromere protein Q
Gene Symbols CENPQ
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 131-268 of human CENPQ (NP_060602.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55166
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.