Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPT Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128698JF646
Description
CENPT Polyclonal antibody specifically detects CENPT in Human samples. It is validated for Western BlotSpecifications
CENPT | |
Polyclonal | |
Western Blot | |
C16orf56, CENP-Tchromosome 16 open reading frame 56, centromere protein T, FLJ13111, FLJ43376, ICEN22, Interphase centromere complex protein 22 | |
The immunogen recognized by this antibody maps to a region between residue 520 and 561 of human centromere protein T using the numbering given in entry NP_079358.3 (GeneID 80152). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
80152 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction