Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563512 Supplier Invitrogen™ Supplier No. PA563512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLHDGVEEAE KKMEEEGVSV SEMEATGAQG PSRVEEAEGH TEVTEAEGSQ GTAEADGPGA SSGDEDASGR AASPESASST PESLQARRHH QFLEP Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 34%.

Component of the CENPA-NAC (nucleosome-associated) complex, a complex that plays a central role in assembly of kinetochore proteins, mitotic progression and chromosome segregation. The CENPA-NAC complex recruits the CENPA-CAD (nucleosome distal) complex and may be involved in incorporation of newly synthesized CENPA into centromeres. Part of a nucleosome-associated complex that binds specifically to histone H3-containing nucleosomes at the centromere, as opposed to nucleosomes containing CENPA. Component of the heterotetrameric CENP-T-W-S-X complex that binds and supercoils DNA, and plays an important role in kinetochore assembly. CENPT has a fundamental role in kinetochore assembly and function. It is one of the inner kinetochore proteins, with most further proteins binding downstream. Required for normal chromosome organization and normal progress through mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPT
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CENPT
Gene Accession No. Q96BT3
Gene Alias C16orf56; CENPT; CENP-T; Centromere protein T; G630055P03Rik; ICEN22; Interphase centromere complex protein 22; RGD1309383
Gene Symbols CENPT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CENPT. Recombinant protein control fragment (Product #RP-98620).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80152
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.