Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CENPT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583540
Description
Immunogen sequence: RGRSHGARSV GRSAHIQASG HLEEQTPRTL LKNILLTAPE SSILMPESVV KPVPAPQAVQ PSRQESSCGS LELQLPELEP PTTLAPGLLA PGRRKQRLRL SVFQQGV.
Component of the CENPA-NAC (nucleosome-associated) complex, a complex that plays a central role in assembly of kinetochore proteins, mitotic progression and chromosome segregation. The CENPA-NAC complex recruits the CENPA-CAD (nucleosome distal) complex and may be involved in incorporation of newly synthesized CENPA into centromeres. Part of a nucleosome-associated complex that binds specifically to histone H3-containing nucleosomes at the centromere, as opposed to nucleosomes containing CENPA. Component of the heterotetrameric CENP-T-W-S-X complex that binds and supercoils DNA, and plays an important role in kinetochore assembly. CENPT has a fundamental role in kinetochore assembly and function. It is one of the inner kinetochore proteins, with most further proteins binding downstream. Required for normal chromosome organization and normal progress through mitosis.
Specifications
CENPT | |
Polyclonal | |
Unconjugated | |
CENPT | |
C16orf56; CENPT; CENP-T; Centromere protein T; G630055P03Rik; ICEN22; Interphase centromere complex protein 22; RGD1309383 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80152 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BT3 | |
CENPT | |
Recombinant protein corresponding to Human CENPT. Recombinant protein control fragment (Product #RP-104730). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction