Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPU Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554562 Supplier Invitrogen™ Supplier No. PA554562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EETYETFDPP LHSTAIYADE EEFSKHCGLS LSSTPPGKEA KRSSDTSGNE ASEIESVKIS AKKPGRKLRP ISDDSESIEE SDTRRKVKSA EKISTQRHEV IRTTASSELS EKPAESV Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 53%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPU
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cenpu
Gene Accession No. Q71F23
Gene Alias 1700029A22Rik; CENP50; CENP-50; Cenpu; CENP-U; CENPU50; Centromere protein of 50 kDa; centromere protein U; ICEN24; Interphase centromere complex protein 24; KLIP1; KSHV latent nuclear antigen interacting protein 1; KSHV latent nuclear antigen-interacting protein 1; LOC617866 protein; MLF1 interacting protein; MLF1-interacting protein; MLF1IP; myeloid leukemia factor 1 interacting protein; PBIP1; Polo-box-interacting protein 1
Gene Symbols Cenpu
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CENPU. Recombinant protein control fragment (Product #RP-88772).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79682
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.