Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPV Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560049
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560049 Supplier Invitrogen™ Supplier No. PA560049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLDLGEQRER WETFQKRQKL TSEGAAKLLL DTFEYQGLVK HTGGCHCGAV RFEVWASADL HIFDCNCSIC KKKQN Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 71%.

CENPV orthologs were detected in a range of species, including plants, but the proline-rich region was detected only in primates, rodents, birds, and amphibians, and long proline stretches were found only in primates. Northern blot analysis showed ubiquitous Cenpv expression in mouse tissues, with highest expression in testis, followed by small intestine, brain, and kidney. Immunofluorescence microscopy of mouse fibroblasts revealed colocalization of Cenpv with stabilized cytoplasmic microtubules. Western blot analysis of Ramos B cells detectedCENPV at an apparent molecular mass of 40 kD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPV
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CENPV
Gene Accession No. Q7Z7K6
Gene Alias 3110013H01Rik; AA671303; Cenpv; CENP-V; Centromere protein V; nuclear protein p30; p30; proline rich 6; proline-rich polypeptide 6; proline-rich protein 6; Prr6; RGD1565681
Gene Symbols CENPV
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CENPV. Recombinant protein control fragment (Product #RP-99873).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201161
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.