Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPW Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566908 Supplier Thermo Scientific Supplier No. PA566908
Add to Cart
Add to Cart

CENPW Polyclonal Antibody

CENPW Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPW
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CENPW
Gene Accession No. Q5EE01
Gene Alias 2610036L11Rik; C6orf173; cancer-upregulated gene 2; cancer-up-regulated gene 2 protein; CANP-W; Cenpw; CENP-W; Centromere protein W; Cug2
Gene Symbols CENPW
Host Species Rabbit
Immunogen Recombinant Human CENPW. Recombinant protein control fragment (Product #RP-107560).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387103
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.