Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Centaurin alpha-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552900
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552900 Supplier Invitrogen™ Supplier No. PA552900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGVFICLSCS GIHRNIPQVS KVKSVRLDAW EEAQVEFMAS HGNDAARARF ESKVPSFYYR PTPSDCQLLR EQWIRAKYER QEFIYPEKQE PYSAGYREGF LWKRG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.

Centaurin alpha-1 (ADAP1, ArfGAP With Dual PH Domains 1) is a Protein Coding gene. Among its related pathways are B Cell Receptor Signaling Pathway (sino) and Arf6 signaling events. Gene Ontology (GO) annotations related to this gene include GTPase activator activity and inositol 1,3,4,5 tetrakisphosphate binding. An important paralog of this gene is ADAP2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Centaurin alpha-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADAP1
Gene Accession No. O75689
Gene Alias 4930431P11Rik; ADAP1; Arf-GAP with dual PH domain-containing protein 1; ArfGAP with dual PH domains 1; CENTA1; centaurin, alpha 1; centaurin-alpha; centaurin-alpha-1; Cnt-a1; GC1L; GCS1L; inositol(1,3,4,5)tetrakisphosphate receptor; p42IP4; phosphatidylinositol-3,4,5-triphosphate binding protein; putative MAPK-activating protein PM25
Gene Symbols ADAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Centaurin alpha-1. Recombinant protein control fragment (Product #RP-89650).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11033
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.