Learn More
Invitrogen™ Centaurin alpha-2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589399
Description
Immunogen sequence: MGDRERNKKR LLELLRAPDT GNAHCADCGA ADPDWASYKL GIFICLNCCG VHRNFPDISR VKSVRLDFWD DSIVEFMIHN GNLRVKAKFE ARVPAFYYIP QANDCLVLKE QWIRAKYERR EFMADGETIS LPGNREGFLW KRGRDNSQFL RRKFVLLARE GLLKYFTKEQ GKSPKAVISI KDLNATFQTE KIGHPHGLQI TYRRDGHTRN LFVYHESGKE IVDWFNALRA ARLQYLKMAF PELPESELVP FLTRNYLKQG FMEKTGPKQK EPFKKRWFAL DCHERRLLYY KNPLDAFEQG QVFLGNKEQG YEAYEDLPKG IRGNRWKAGL TIVTPERRFV LTCPSEKEQQ EWLESLRGVL SSPLTPLNRL TASTESGRSS R; Positive Samples: 293T; Cellular Location: Cell membrane, Cytoplasm.
GTPase-activating protein for the ADP ribosylation factor family (Potential). Binds phosphatidylinositol 3, 4, 5-trisphosphate (PtdInsP3) and inositol 1, 3, 4, 5-tetrakisphosphate (InsP4). Possesses a stoichiometry of two binding sites for InsP4 with identical affinity.
Specifications
Centaurin alpha-2 | |
Polyclonal | |
Unconjugated | |
ADAP2 | |
Adap2; Arf-GAP with dual PH domain-containing protein 2; ArfGAP with dual PH domains 2; CENTA2; centaurin; centaurin beta; centaurin, alpha 2; centaurin-alpha 2 protein; centaurin-alpha-2; Centaurin-alpha2 protein; cent-b; cnt-a2; HSA272195 | |
Rabbit | |
Affinity chromatography | |
RUO | |
55803 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.43 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9NPF8 | |
ADAP2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-381 of human ADAP2 (NP_0608741). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.