Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Centaurin alpha-2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518207
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
GTPase-activating protein for the ADP ribosylation factor family (Potential). Binds phosphatidylinositol 3, 4, 5-trisphosphate (PtdInsP3) and inositol 1, 3, 4, 5-tetrakisphosphate (InsP4). Possesses a stoichiometry of two binding sites for InsP4 with identical affinity.
Specifications
Centaurin alpha-2 | |
Polyclonal | |
Unconjugated | |
ADAP2 | |
Adap2; Arf-GAP with dual PH domain-containing protein 2; ArfGAP with dual PH domains 2; CENTA2; centaurin; centaurin beta; centaurin, alpha 2; centaurin-alpha 2 protein; centaurin-alpha-2; Centaurin-alpha2 protein; cent-b; cnt-a2; HSA272195 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
55803 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9NPF8 | |
ADAP2 | |
Synthetic peptide sequence (RLTASTESGRSSR) corresponding to the C-terminus amino acids of ADAP2 (aa 369-381). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction