Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Centaurin gamma 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986916
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6916 Supplier Bioss Supplier No. BS5930R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene belongs to the centaurin gamma-like family. It mediates anti-apoptotic effects of nerve growth factor by activating nuclear phosphoinositide 3-kinase. It is overexpressed in cancer cells, and promotes cancer cell invasion. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.

Specifications

Antigen Centaurin gamma 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Agap2
Gene Accession No. Q99490
Gene Alias AGAP2; AGAP-2; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 2; ArfGAP with GTPase domain, ankyrin repeat and PH domain 2; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2; centaurin gamma1; centaurin, gamma 1; Centaurin-gamma-1; Centg1; Cnt-g1; FLJ16430; GGAP2; GTP-binding and GTPase activating protein 2; GTP-binding and GTPase-activating protein 2; Kiaa0167; mKIAA0167; nuclear GTPase PIKE; Phosphatidylinositol 3-kinase enhancer; phosphatidylinositol-3-kinase enhancer; phosphoinositide 3-kinase enhancer; PIKE
Gene Symbols Agap2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Centaurin gamma 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116986
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.