Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CENTB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584987
Description
Immunogen sequence: QGHEELSRLS QYRKELGAQL HQLVLNSARE KRDMEQRHVL LKQKELGGEE PEPSLREGPG GLVMEGHLFK.
ACAP1, also known as Centaurin-beta-1 (CENTB1), is a 740 amino acid containing transport carrier belonging to the Centaurin family with three ANK repeats, an Arf-GAP domain, a BAR domain and a PH domain that binds to phospholipids. ACAP1, a GTPase-activating protein (GAP) for ADP ribosylation factor 6 (ARF6), is required for clathrin-dependent export of proteins from recycling endosomes to trans-Golgi network and cell surface. It mediates CENTB1-binding to phosphatidylinositol 3,5-bisphosphate (PIP2) or phosphatidylinositol 3,4,5-trisphosphate (PIP3) containing membranes. Along with ACAP2, ACAP1 is recruited to platelet-derived growth factor (PDGF) induced dorsal membrane ruffles in NIH 3T3 mouse fibroblasts, while over expression inhibited ruffle formation. It is widely expressed in most tissues with high expression in lung and spleen and low expression in heart, kidney, liver and pancreas.
Specifications
CENTB1 | |
Polyclonal | |
Unconjugated | |
ACAP1 | |
Acap1; Arf GAP with coiled coil, ANK repeat and PH domains 1; arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 1; ArfGAP with coiled-coil, ankyrin repeat and PH domains 1; centaurin beta 1; centaurin, beta 1; centaurin-beta-1; Centb1; cnt-b1; KIAA0050 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9744 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15027 | |
ACAP1 | |
Recombinant protein corresponding to Human ACAP1. Recombinant protein control fragment (Product #RP-107134). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction