Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENTD1 Antibody, Novus Biologicals™
SDP

Catalog No. p-5054923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224620SS 0.025 mg
NBP224620 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224620SS Supplier Novus Biologicals Supplier No. NBP224620SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CENTD1 Polyclonal specifically detects CENTD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CENTD1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1.0-2.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias Arf and Rho GAP adapter protein 2, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2, centaurin, delta 1, Centaurin-delta-1, CENTD1arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 2, cnt-d1, FLJ13675, FLJ44916, KIAA0580centaurin-delta-1, PARX, Protein PARX
Gene Symbols ARAP2
Host Species Rabbit
Immunogen A portion of amino acids 1670-1704 of human ARAP2 was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116984
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_c, CRA_d and CRA_e), 93% homologous in rhesus monkey, 87% homologous in mouse and rat, 81% homologous in dog and 75% homologous in opossum.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.