Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Checkers Center 3-Channel Rubber Duct Office Cable Protector
Click to view available options
Length (English):
5 ft.
10 ft.
Description
- Center Channel Sized To Fit Standard Power Cords And CAT5 Cable
- Protector Must Be Slit For Standard Power Cords Due To Size Of Plug Connectors (Supplied Unslit)
- Reduces Tripping Hazards
- Protects And Organizes Portable Cords And Cables
- Non-Metallic, Industrial-Grade Rubber Construction
- Multiple Sizes For Various Cable Diameters
- Grooved Underside For Better Traction
- Optional Connector Kit Available For Most Sizes
Specifications
Specifications
Length (English) | 5 ft. |
Color | Black |
Material | Rubber |
Product Type | Cable Protector |
Length (Metric) | 1.5 m |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction