Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CENTG1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519119
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
The protein encoded by this gene belongs to the centaurin gamma-like family. It mediates anti-apoptotic effects of nerve growth factor by activating nuclear phosphoinositide 3-kinase. It is overexpressed in cancer cells, and promotes cancer cell invasion. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| CENTG1 | |
| Polyclonal | |
| Unconjugated | |
| Agap2 | |
| AGAP2; AGAP-2; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 2; ArfGAP with GTPase domain, ankyrin repeat and PH domain 2; arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2; centaurin gamma1; centaurin, gamma 1; centaurin-gamma-1; CENTG1; cnt-g1; FLJ16430; GGAP2; GTP-binding and GTPase activating protein 2; GTP-binding and GTPase-activating protein 2; KIAA0167; mKIAA0167; nuclear GTPase PIKE; phosphatidylinositol 3-kinase enhancer; phosphatidylinositol-3-kinase enhancer; phosphoinositide 3-kinase enhancer; Pike | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 116986 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q99490 | |
| Agap2 | |
| Synthetic peptide sequence (QASLDSIREAVINSQ) corresponding to the internal amino acids of AGAP2 (aa 392-397). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction